Anti-SV40 T-antigen Mouse monoclonal antibody [clone: PAb419]

ABCAAB16848-1EA 588 EUR
ABCAAB16848-1
Anti-SV40 T-antigen Mouse monoclonal antibody [clone: PAb419]
Type: Primary
Antigen: SV40 T-antigen
Clonality: monoclonal
Clone: PAb419
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR