Anti-KIT Rabbit polyclonal antibody

ABCAAB16832-100EA 588 EUR
ABCAAB16832-100
Anti-KIT Rabbit polyclonal antibody
Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR