Anti-ITGA2B/B3 Mouse monoclonal antibody [clone: 290.5C10]

ABCAAB16822-50EA 588 EUR
ABCAAB16822-50
Anti-ITGA2B/B3 Mouse monoclonal antibody [clone: 290.5C10]
Type: Primary
Antigen: ITGA2B/B3 (integrin, alpha 2B/beta 3)
Clonality: monoclonal
Clone: 290.5C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR