Anti-APC Mouse Monoclonal Antibody [clone: CC-1]

Proveedor: Abcam
ABCAAB16794-50EA 839 EUR
ABCAAB16794-50
Anti-APC Mouse Monoclonal Antibody [clone: CC-1]
Anticuerpos
Type: Primary
Antigen: APC (adenomatous polyposis coli)
Clonality: monoclonal
Clone: CC-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR