Anti-BRCA1 Mouse monoclonal antibody unconjugated [clone: MS110]
Proveedor: Abcam
ABCAAB16780-100EA 585 EUR
ABCAAB16780-100
Anti-BRCA1 Mouse monoclonal antibody unconjugated [clone: MS110]
Anticuerpos
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: MS110
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human