Anti-GPX1 Mouse monoclonal antibody [clone: 42C9]

ABCAAB16736-100EA 645 EUR
ABCAAB16736-100
Anti-GPX1 Mouse monoclonal antibody [clone: 42C9]
Type: Primary
Antigen: GPX1 (glutathione peroxidase 1)
Clonality: monoclonal
Clone: 42C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR