Anti-GLRX2 Mouse monoclonal antibody [clone: 1A3]

ABCAAB16733-100EA 588 EUR
ABCAAB16733-100
Anti-GLRX2 Mouse monoclonal antibody [clone: 1A3]
Type: Primary
Antigen: GLRX2 (glutaredoxin 2)
Clonality: monoclonal
Clone: 1A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR