Anti-BAFF Receptor Mouse Monoclonal Antibody (Biotin) [clone: 11C1]

Proveedor: Abcam
ABCAAB16582-100EA 781 EUR
ABCAAB16582-100
Anti-BAFF Receptor Mouse Monoclonal Antibody (Biotin) [clone: 11C1]
Anticuerpos
Type: Primary
Antigen: BAFF Receptor
Clonality: monoclonal
Clone: 11C1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR