Anti-PPP4C Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB16475-100EA 773 EUR
ABCAAB16475-100
Anti-PPP4C Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PPP4C (protein phosphatase 4, catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR