Anti-PSEN2 Chicken polyclonal antibody

ABCAAB16245-100EA 611 EUR
ABCAAB16245-100
Anti-PSEN2 Chicken polyclonal antibody
Type: Primary
Antigen: PSEN2 (presenilin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR