Anti-LEP Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB16227-50EA 595 EUR
ABCAAB16227-50
Anti-LEP Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LEP (leptin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR