Anti-COX2 / Cyclooxygenase 2 Mouse monoclonal antibody [clone: AS73]

ABCAAB16118-50EA 611 EUR
ABCAAB16118-50
Anti-COX2 / Cyclooxygenase 2 Mouse monoclonal antibody [clone: AS73]
Type: Primary
Antigen: COX2 / Cyclooxygenase 2
Clonality: monoclonal
Clone: AS73
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR