Anti-NLRP1 Mouse Monoclonal Antibody [clone: Nalpy1-4]

Proveedor: Abcam
ABCAAB16091-50EA 745 EUR
ABCAAB16091-50
Anti-NLRP1 Mouse Monoclonal Antibody [clone: Nalpy1-4]
Anticuerpos
Type: Primary
Antigen: NLRP1 (NLR family, pyrin domain containing 1)
Clonality: monoclonal
Clone: Nalpy1-4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR