Anti-MAGE 1 Rabbit polyclonal antibody

ABCAAB16034-7EA 611 EUR
ABCAAB16034-7
Anti-MAGE 1 Rabbit polyclonal antibody
Type: Primary
Antigen: MAGE 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR