Anti-SMYD3 Rabbit polyclonal antibody

ABCAAB16027-100EA 0 EUR
ABCAAB16027-100
Anti-SMYD3 Rabbit polyclonal antibody
Type: Primary
Antigen: SMYD3 (SET and MYND domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR