Anti-Streptococcus Enterotoxin B Rabbit polyclonal antibody

ABCAAB16022-500EA 604 EUR
ABCAAB16022-500
Anti-Streptococcus Enterotoxin B Rabbit polyclonal antibody
Type: Primary
Antigen: Streptococcus Enterotoxin B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR