Anti-Staphylococcus Enterotoxin A Sheep polyclonal antibody

ABCAAB15918-500EA 604 EUR
ABCAAB15918-500
Anti-Staphylococcus Enterotoxin A Sheep polyclonal antibody
Type: Primary
Antigen: Staphylococcus Enterotoxin A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR