Anti-KCNC2 Chicken Polyclonal Antibody

Proveedor: Abcam
ABCAAB15869-50EA 840 EUR
ABCAAB15869-50
Anti-KCNC2 Chicken Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: KCNC2 (potassium voltage-gated channel, Shaw-related subfamily, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR