Anti-Coronin 3 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB15719-100EA 0 EUR
ABCAAB15719-100
Anti-Coronin 3 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Coronin 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR