Anti-EML2 Mouse monoclonal antibody unconjugated [clone: 546-2]

Proveedor: Abcam
ABCAAB15693-100EA 0 EUR
ABCAAB15693-100
Anti-EML2 Mouse monoclonal antibody unconjugated [clone: 546-2]
Anticuerpos
Type: Primary
Antigen: EML2 (echinoderm microtubule associated protein like 2)
Clonality: monoclonal
Clone: 546-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR