Anti-MHC class I Rat Monoclonal Antibody [clone: ERMP42]
Proveedor: Abcam
ABCAAB15680-50EA 0 EUR
ABCAAB15680-50
Anti-MHC class I Rat Monoclonal Antibody [clone: ERMP42]
Anticuerpos
Type: Primary
Antigen: MHC class I
Clonality: monoclonal
Clone: ERMP42
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse