Anti-CD163 Mouse monoclonal antibody Biotin [clone: RM3/1]

ABCAAB15676-50EA 310 EUR
ABCAAB15676-50
Anti-CD163 Mouse monoclonal antibody Biotin [clone: RM3/1]
Type: Primary
Antigen: CD163 (CD163 molecule)
Clonality: monoclonal
Clone: RM3/1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR