Anti-pan Macrophage Mouse Monoclonal Antibody [clone: RM-1]

Proveedor: Abcam
ABCAAB15615-50EA 0 EUR
ABCAAB15615-50
Anti-pan Macrophage Mouse Monoclonal Antibody [clone: RM-1]
Anticuerpos
Type: Primary
Antigen: pan Macrophage
Clonality: monoclonal
Clone: RM-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR