Anti-PROCR Rat Monoclonal Antibody [clone: LMR-42]

Proveedor: Abcam
ABCAAB15604-500EA 739 EUR
ABCAAB15604-500
Anti-PROCR Rat Monoclonal Antibody [clone: LMR-42]
Anticuerpos
Type: Primary
Antigen: PROCR (protein C receptor, endothelial)
Clonality: monoclonal
Clone: LMR-42
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR