Anti-MKI67 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB15580-100EA 635 EUR
ABCAAB15580-100
Anti-MKI67 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now