Anti-MUC1 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB15481-500EA 837 EUR
ABCAAB15481-500
Anti-MUC1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR