Anti-MMP15 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB15475-500EA 997 EUR
ABCAAB15475-500
Anti-MMP15 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MMP15 (matrix metallopeptidase 15 (membrane-inserted))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR