Anti-ITGB5 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB15459-500EA 838 EUR
ABCAAB15459-500
Anti-ITGB5 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ITGB5 (integrin, beta 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR