Anti-ACE2 Rabbit polyclonal antibody

ABCAAB15347-100EA 626 EUR
ABCAAB15347-100
Anti-ACE2 Rabbit polyclonal antibody
Type: Primary
Antigen: ACE2 (angiotensin I converting enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR