Anti-MSLN Mouse monoclonal antibody [clone: SPM143]

ABCAAB15231-7EA 611 EUR
ABCAAB15231-7
Anti-MSLN Mouse monoclonal antibody [clone: SPM143]
Type: Primary
Antigen: MSLN (mesothelin)
Clonality: monoclonal
Clone: SPM143
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR