Anti-Caffeine Mouse monoclonal antibody [clone: 8.F.28]

ABCAAB15219-500EA 604 EUR
ABCAAB15219-500
Anti-Caffeine Mouse monoclonal antibody [clone: 8.F.28]
Type: Primary
Antigen: Caffeine
Clonality: monoclonal
Clone: 8.F.28
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR