Anti-BZE Mouse monoclonal antibody [clone: 1.BB.868]

ABCAAB15216-500EA 604 EUR
ABCAAB15216-500
Anti-BZE Mouse monoclonal antibody [clone: 1.BB.868]
Type: Primary
Antigen: BZE (Benzoylecgonine)
Clonality: monoclonal
Clone: 1.BB.868
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR