Anti-D4 GDI Rabbit polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB15198-500EA 753 EUR
ABCAAB15198-500
Anti-D4 GDI Rabbit polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: D4 GDI
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR