Anti-AQP5 Rabbit polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB15119-25EA 739 EUR
ABCAAB15119-25
Anti-AQP5 Rabbit polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: AQP5 (aquaporin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR