Anti-CEBPA Rabbit polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB15048-50EA 0 EUR
ABCAAB15048-50
Anti-CEBPA Rabbit polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: CEBPA (CCAAT/enhancer binding protein (C/EBP), alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR