Anti-PICK1 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB15001-50EA 739 EUR
ABCAAB15001-50
Anti-PICK1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PICK1 (protein interacting with PRKCA 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR