Anti-H3F3AP6 Rabbit polyclonal antibody

ABCAAB14975-50EA 645 EUR
ABCAAB14975-50
Anti-H3F3AP6 Rabbit polyclonal antibody
Type: Primary
Antigen: H3F3AP6 (H3 histone, family 3A, pseudogene 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR