Anti-ATP5J Mouse monoclonal antibody unconjugated [clone: 15H4C4]

Proveedor: Abcam
ABCAAB14748-100EA 855 EUR
ABCAAB14748-100
Anti-ATP5J Mouse monoclonal antibody unconjugated [clone: 15H4C4]
Anticuerpos
Type: Primary
Antigen: ATP5J (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F6)
Clonality: monoclonal
Clone: 15H4C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR