Anti-STAT1 Mouse Monoclonal Antibody [clone: SM2]

Proveedor: Abcam
ABCAAB14743-100EA 739 EUR
ABCAAB14743-100
Anti-STAT1 Mouse Monoclonal Antibody [clone: SM2]
Anticuerpos
Type: Primary
Antigen: STAT1 (signal transducer and activator of transcription 1, 91kDa)
Clonality: monoclonal
Clone: SM2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR