Anti-proBNP Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 13G12]

ABCAAB14712-200EA 654 EUR
ABCAAB14712-200
Anti-proBNP Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 13G12]
Type: Primary
Antigen: proBNP
Clonality: monoclonal
Clone: 13G12
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR