Anti-FOXP3 Goat polyclonal antibody

ABCAAB14551-100EA 577 EUR
ABCAAB14551-100
Anti-FOXP3 Goat polyclonal antibody
Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR