Anti-KSR1 Mouse monoclonal antibody

ABCAAB14516-50EA 611 EUR
ABCAAB14516-50
Anti-KSR1 Mouse monoclonal antibody
Type: Primary
Antigen: KSR1 (kinase suppressor of ras 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR