Anti-GHRL Rabbit polyclonal antibody

ABCAAB14480-500EA 577 EUR
ABCAAB14480-500
Anti-GHRL Rabbit polyclonal antibody
Type: Primary
Antigen: GHRL (ghrelin/obestatin prepropeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR