Anti-FZD5 Rabbit polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB14475-50EA 805 EUR
ABCAAB14475-50
Anti-FZD5 Rabbit polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: FZD5 (frizzled class receptor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR