Anti-DEFB4A Mouse monoclonal antibody unconjugated [clone: B 235-I]

Proveedor: Abcam
ABCAAB14426-50EA 0 EUR
ABCAAB14426-50
Anti-DEFB4A Mouse monoclonal antibody unconjugated [clone: B 235-I]
Anticuerpos
Type: Primary
Antigen: DEFB4A (defensin, beta 4A)
Clonality: monoclonal
Clone: B 235-I
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR