Anti-Smt3 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB14405-500EA 771 EUR
ABCAAB14405-500
Anti-Smt3 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Smt3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR