Anti-RLN1 Mouse monoclonal antibody [clone: willi]

ABCAAB14326-50EA 604 EUR
ABCAAB14326-50
Anti-RLN1 Mouse monoclonal antibody [clone: willi]
Type: Primary
Antigen: RLN1 (relaxin 1)
Clonality: monoclonal
Clone: willi
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR