Anti-PTPN21 Chicken polyclonal antibody

ABCAAB14315-50EA 686 EUR
ABCAAB14315-50
Anti-PTPN21 Chicken polyclonal antibody
Type: Primary
Antigen: PTPN21 (protein tyrosine phosphatase, non-receptor type 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR