Anti-CGB5 Chicken polyclonal antibody

ABCAAB14301-50EA 653 EUR
ABCAAB14301-50
Anti-CGB5 Chicken polyclonal antibody
Type: Primary
Antigen: CGB5 (chorionic gonadotropin, beta polypeptide 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR