Anti-E2F2 Chicken polyclonal antibody
ABCAAB14292-50EA 619 EUR
ABCAAB14292-50
Anti-E2F2 Chicken polyclonal antibody
Type: Primary
Antigen: E2F2 (E2F transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse